Ammonium-1-pyrrolidincarbodithioat, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD
548000-100
548000-100EA
87
EUR
548000-100
Ammonium-1-pyrrolidincarbodithioat, Millipore®
Ammonium-1-pyrrolidincarbodithioat
Formel:
C₅H₁₂N₂S₂ Molecular Weight: 146.3 · 18.0 g/mol |
MDL:
MFCD00012720 CAS-Nummer: 5108-96-3 EINECS: 225-834-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...