Mikrovolumenspritzen 5 µl
Lieferant: Trajan Scientific and Medical
001000 001050 001100 001400 001450 001495 001450CAL
549-0267EA 56.4 EUR
549-0267 549-0268 549-0269 549-0272 549-0273 549-0439 SGEA001450CAL
Mikrovolumenspritzen 5 µl
Spritzen Spritzen für die Chromatographie
These are plunger-in-barrel syringes. Each plunger is individually fitted for each microsyringe for perfect sealing.
  • Longer life
  • Superior performance and robustness
  • Reduced carryover
Plunger protection incorporates a plunger protection guide and plunger button reinforcement to minimise the risk of bent plungers.
Guided plunger microvolume syringes have an extended barrel and reinforced plunger that provides additional protection.
* SuperfleX, mit flexiblem Kolben. SuperfleX microvolume syringes use a super elastic alloy plunger that will not bend or kink.