4,5-Dibromobenzene-1,2-diol 90%, technische Qualität, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
550248-5G
550248-5GEA
248
EUR
550248-5G
4,5-Dibromobenzene-1,2-diol 90%, technische Qualität, Sigma-Aldrich®
4,5-Dibromobenzene-1,2-diol
Formel:
C₆H₄Br₂O₂ Schmelzpunkt: 111,..116 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00233157 CAS-Nummer: 2563-26-0 |
Spezifikation Testergebnisse
Appearance (Colour); White to Grey | Conforms to Requirements |
Appearance (Form); Powder, Crystals, Crystalline Powder and/or Chunks | Conforms to Requirements |
Infrared spectrum | Conforms to Structure |
Proton NMR spectrum | Conforms to Structure |
Carbon | 23.7 - 30.1 % |
Purity (GC) | ≥88.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...