Wasserstoffperoxid 30%, Finyte® zur mikroelektronischen Verwendung, J.T.Baker®

Lieferant: Avantor
Gefahr


Veränderungsmanagement-Kategorie= R

5516-07EA 940 EUR
5516-07
Wasserstoffperoxid 30%, Finyte® zur mikroelektronischen Verwendung, J.T.Baker®
Wasserstoffperoxid
Formel: H₂O₂
MDL: MFCD00011333
CAS-Nummer: 7722-84-1
UN: 2014
ADR: 5.1,II

Bestellen

Spezifikation Testergebnisse

For Microelectronic Use
Assay (H₂O₂) 30.0 - 32.0 %
Color (APHA) ≤ 6
Free Acid (µeq/g) ≤ 0.6
Residue after Evaporation ≤ 5 ppm
Chloride (Cl) ≤ 0.06 ppm
Nitrate (NO₃) ≤ 0.4 ppm
Phosphate (PO₄) ≤ 0.20 ppm
Sulfate (SO₄) ≤ 0.2 ppm
Trace Impurities - Aluminum (Al) ≤ 10.0 ppb
Arsenic and Antimony (as As) ≤ 5.0 ppb
Trace Impurities - Barium (Ba) ≤ 10.0 ppb
Trace Impurities - Beryllium (Be) ≤ 10.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 10.0 ppb
Trace Impurities - Boron (B) ≤ 10.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 10.0 ppb
Trace Impurities - Calcium (Ca) ≤ 10.0 ppb
Trace Impurities - Chromium (Cr) ≤ 10.0 ppb
Trace Impurities - Cobalt (Co) ≤ 10.0 ppb
Trace Impurities - Copper (Cu) ≤ 10.0 ppb
Trace Impurities - Gallium (Ga) ≤ 10.0 ppb
Trace Impurities - Germanium (Ge) ≤ 10.0 ppb
Trace Impurities - Gold (Au) ≤ 10.0 ppb
Trace Impurities - Iron (Fe) ≤ 10.0 ppb
Trace Impurities - Lead (Pb) ≤ 10.0 ppb
Trace Impurities - Lithium (Li) ≤ 10.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 10.0 ppb
Trace Impurities - Manganese (Mn) ≤ 10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤ 10.0 ppb
Trace Impurities - Nickel (Ni) ≤ 10.0 ppb
Trace Impurities - Niobium (Nb) ≤ 10.0 ppb
Trace Impurities - Potassium (K) ≤ 10.0 ppb
Trace Impurities - Silicon (Si) ≤ 10.0 ppb
Trace Impurities - Silver (Ag) ≤ 10.0 ppb
Trace Impurities - Sodium (Na) ≤ 10.0 ppb
Trace Impurities - Strontium (Sr) ≤ 10.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 10.0 ppb
Trace Impurities - Thallium (Tl) ≤ 10.0 ppb
Trace Impurities - Tin (Sn) ≤ 10.0 ppb
Trace Impurities - Titanium (Ti) ≤ 10.0 ppb
Trace Impurities - Vanadium (V) ≤ 10.0 ppb
Trace Impurities - Zinc (Zn) ≤ 10.0 ppb
Trace Impurities - Zirconium (Zr) ≤ 10.0 ppb
Particle Count - 0.2 µm and greater ≤ 1175 par/ml
Particle Count - 0.5 µm and greater, For Information Only For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR