TLC plates, special layers, polyester sheets, IONEX, POLYGRAM®
Lieferant: MACHEREY-NAGEL
806013 806023
552-0165EA 228 EUR
552-0165 552-0166
TLC plates, special layers, polyester sheets, IONEX, POLYGRAM®
TLC-Platten
These are special TLC layers of silica mixed with ion exchange resins available as two different layers, both containing an inert organic binder.
  • IONEX-25 SA-Na: mixture of silica and a strongly acidic cation exchanger coated to polyester sheets
  • IONEX-25 SB-AC: mixture of silica and a strongly basic anion exchanger coated to polyester sheets
They are recommended for amino acids, e.g., in protein and peptide hydrolysates, in seeds and fodder, in biological fluids; for racemate separation in peptide syntheses, for the separation of nucleic acidMore Product Information