TLC plates, mixed layers, glass backing
Lieferant: MACHEREY-NAGEL
810043
552-0217EA 274 EUR
552-0217
TLC plates, mixed layers, glass backing
TLC-Platten
Glass plates with mixed layers for TLC.
Alox/CEL-AC-Mix-25: mixed layer of aluminium oxide G and acetylated cellulose recommended for separation of PAHs.
SILCEL-Mix-25: mixed layer of cellulose and silica, recommended for separation of preservatives and other antimicrobial compounds.