HPTLC plates, nano silica gel 60, special layer for PAH analysis, Nano-SIL PAH
Lieferant: MACHEREY-NAGEL
811051
552-0234EA 545 EUR
552-0234
HPTLC plates, nano silica gel 60, special layer for PAH analysis, Nano-SIL PAH
TLC-Platten
Glass plates with a special special HPTLC silica layer for PAH analysis.
  • Nano silica gel (particle size 2 to 10 μm)
  • Caffeine impregantion
  • Recommended for PAH analysis
Recommended application: determination of the six PAHs according to German drinking water specifications (TVO) in accordance with German standard DIN 38407 part 7.
Base material: Nano silica 60, specific surface (BET) ~500 m²/g, specific pore volume 0,75 ml/g, particle size 2 to 10 μm
Impregnation: Caffeine