TLC developing chambers
Lieferant: MACHEREY-NAGEL
814021
552-0273EA 40.4 EUR
552-0273
TLC developing chambers
TLC-Entwicklungskammern
Mason jar shaped developing chambers for TLC application. Glass of hydrolytic category III, lid is made of PP with LDPE sealing disk.
Recommended TLC plate size: Up to 4×8 cm