HPTLC plates, silica gel 60, unmodified layers, Nano-ADAMANT
Lieferant: MACHEREY-NAGEL
821110 821120 821140 821150
552-0344EA 190 EUR
552-0344 552-0345 552-0347 552-0348
HPTLC plates, silica gel 60, unmodified layers, Nano-ADAMANT
TLC-Platten
Unmodified HPTLC silica layers available as glass plates with or without fluorescent indicator. The narrow fractionation of the silica particles features theoretical plate heights, which are one order of magnitude smaller than on standard silica layers. Thus, Nano-ADAMANT has the advantage of sharper separations, shorter developing times, and increased detection sensitivity with equal selectivity.
  • Good hardness and abrasion resistance due to an optimized binder system
  • Increased separation efficiency due to an optimised particle size distribution
  • High
More Product Information