Ruthenium ≥99,95% (Metall-Basis), Plätzchen 3-6 mm

Lieferant: Thermo Fisher Scientific

55236.06
55236.06EA 595 EUR
55236.06
Ruthenium ≥99,95% (Metall-Basis), Plätzchen 3-6 mm
Ruthenium

Purity: 99.95% CAS No.: 7440-18-8 Physical Form: ≈1.5-2.5g/pellet Appearance: Grey Odour: Odourless MDL No.: MFCD00011207

Formel: Ru
Lagertemperatur: Raumtemperatur
MDL: MFCD00011207
CAS-Nummer: 7440-18-8
EINECS: 231-127-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR