Cutter for stainless steel tubing, cutting wheel type
Lieferant: VICI JOUR RESEARCH
JR-792
554-1103EA 151 EUR
554-1103
Cutter for stainless steel tubing, cutting wheel type
Schneidewerkzeuge
This is the ideal tool for cutting ¹/₁₆ and ¹/₈" stainless steel tubing, leaving smooth, uniform cuts requiring a minimum of deburring and/or reaming.
The easily replaced cutting wheel scores tubing, leaving cuts which allow tube-to-tube and tube-to-frit connections.