Zweiteilige Fittings, Fingertight, Upchurch Scientific®
Lieferant: UPCHURCH SCIENTIFIC
F-300X F-113 F-142 F-142X F-330X
554-1789EA 170 EUR
554-1789 554-1785 554-4696 554-1788 554-5681
Zweiteilige Fittings, Fingertight, Upchurch Scientific®
Fittings und Verbinder
PEEK, Edelstahl oder PCTFE
Double winged fittings with 10-32 thread, designed to be used with 1/16" OD tubing. They are rated to 6000 psi (414 bar).
F-142x ferrules are designed to be used with 1/16" OD tubing. They are rated to 6000 psi (414 bar). F-113 ferrules are designed to be used with 1/32" OD tubing. They are rated at 3500 - 6000 psi (241 - 414 bar).