Hochdruck-Fittings, Upchurch Scientific®
Lieferant: UPCHURCH SCIENTIFIC
P-742 P-760 P-779 P-779-01 P-704 P-704-01 P-742-01 U-283 U-284 U-287 U-288
554-1803EA 86.9 EUR
554-1803 554-1804 554-1805 554-5689 554-1802 554-5688 554-4728 554-5690 554-5691 554-5692 554-5693
Hochdruck-Fittings, Upchurch Scientific®
Fittings und Verbinder
Fittings ohne Totvolumen aus PEEK: Die Einheiten ohne Totvolumen werden komplett mit zwei F-300 von Hand angezogenen, dichten Anschlüssen für Kapillaren mit einem Außendurchmesser von ¹/₁₆" geliefert und sind für einen Druck von 344 bar (5000 psi) ausgelegt.
PEEK NanoTight™ union: The unions come complete with two F-331N fingertight fittings and are pressure rated to 5000 psi  (344 bar). The internal design of the unions reduces tubing misalignment. When using ¹/₁₆" Ext.Ø tubing sleeves to connect capillary tubing, the webbed thru-hole minimises breaking of fused silica while adding only miniscule swept volume. The results are: Fewer blockages, fewer flow rate reductions and fewer back pressure problems.
PCTFE column coupler: These couplers will connect most columns with 10-32 internal threads to a pre-column filter or guard column with the same threads. They may also be used to join two columns in series.
The unions come complete with two F-331N fingertight fittings and are pressure rated to 5000 psi (344 bar). The internal design of the unions reduces tubing misalignment. When using 1/16" OD tubing sleeves to connect capillary tubing, the webbed thru-hole minimises breaking of fused silica while adding only miniscule swept volume. The results are: