VWR® Hichrom Pliers, for Stainless Steel Tubing
Lieferant: Avantor
554-7744EA 77.8 EUR
554-7744
VWR® Hichrom Pliers, for Stainless Steel Tubing
Zangen
These pliers are designed for cutting 1/16" stainless steel tubing and are especially suitable for tubing of 0,010" and 0,020" ID.
  • Leaves smooth and uniform cuts
  • De-burring may be necessary to achieve an optimum finish