Resveratrol, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol

554325-25
554325-25EA 87 EUR
554325-25
Resveratrol, Millipore®
Resveratrol
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
Schmelzpunkt: 260
Dichte: 1.36 g/cm³ (20 °C)
Lagertemperatur: Gefrierschrank
MDL: MFCD00133799
CAS-Nummer: 501-36-0
Merck Index: 12,08326

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR