Säulenendstopfen
Lieferant: VICI JOUR RESEARCH
JR-4073-10 JR-4072-10 JR-4071-10
555-3595EA 15.1 EUR
555-3595 554-1146 554-0759
Säulenendstopfen
Fittings und Verbinder
These nylon column end plugs fit all columns with 10-32 threads. The long design and threading down to the ferrule ensure safe closing of valuable columns.
  • Available with a 5/16" wrench flat
  • Easy removal