HPLC columns, HyPURITY™ C8
Lieferant: Thermo Fisher Scientific
22205-254630 22205-154630 22205-104630 22205-054630
555-7070EA 1080 EUR
555-7070 555-7064 555-7057 555-7052
HPLC columns, HyPURITY™ C8
Chromatographiesäulen
The unique polar embedded C8 columns can also act to positively repel ionized basic species, resulting in excellent peak shape for these typically problematic compounds.
  • A polar embedded stationary phase for alternate selectivity
  • High quality and highly pure silica columns