HPLC columns, HyPURITY™ C18
Lieferant: Thermo Fisher Scientific
22105-013001 22105-012101 22103-254630 22103-154630 22103-153030 22103-152130 22105-052130 22103-102130 22105-053030 22103-052130 22105-014001 22105-254630 22105-254030 22105-154630 22105-154030 22105-153030 22105-152130 22105-032130 22103-154030 22103-054630 22103-104630 22105-252130 22105-253030 22105-104630 22105-102130 22105-054630 22103-124630
555-7005EA 533 EUR
555-7005 555-7003 555-7000 555-6998 555-6996 555-6995 555-7083 555-6979 555-7084 555-6983 555-7006 555-7105 555-7104 555-7099 555-7098 555-7097 555-7096 555-7078 555-6997 555-6975 555-6982 555-7102 555-7103 555-7091 555-7088 555-7086 555-6992
HPLC columns, HyPURITY™ C18
Chromatographiesäulen
HyPURITY™ C18 columns are ideal for chromatography of large peptides, small molecules, with unbeatable peak shape for all sample types. The homogeneous surface ensures uniform bonding coverage and eliminates silanolanalyte interactions that cause poor peak shapes for bases, acids and chelating compounds.
  • Highly reproducible, rugged and robust
  • Stable at low pH