Capillary columns, TraceGOLD™ TG-WaxMS
Lieferant: Thermo Fisher Scientific
26088-1300 26088-2130 26088-0200 26088-1420 26088-1430 26088-1440 26088-1540 26088-1550 26088-1560 26088-2230 26088-2120 26088-3360 26088-2240 26088-2250 26088-2350 26088-2360 26088-2370 26088-2860 26088-2970 26088-2980 26088-3090 26088-3100
555-8594EA 504 EUR
555-8594 555-8580 555-8595 555-8590 555-8588 555-8586 555-8585 555-8584 555-8583 555-8579 555-8581 555-8625 555-8578 555-8577 555-8576 555-8575 555-8636 555-8632 555-8630 555-8629 555-8628 555-8627
Capillary columns, TraceGOLD™ TG-WaxMS
Chromatographiesäulen
TraceGOLD™ TG-WaxMS columns are manufactured for better column-to-column reproducibility.
  • Polar phase, polyethylene glycol
  • Polar-deactivated surface tightly binds polymer for excellent thermal stability
  • Resists oxidative damage, damage from strongly acidic or basic volatiles better than silicone solid phases
  • Equivalent to USP G14, G15, G16, G20 and G39 phases