4-Chloro-7-chlorosulphonyl-2,1,3-benzoxadiazole 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: 4-Chloro-7-chlorosulphonylbenzofurazan, 4-Chloro-2,1,3-benzoxadiazole-7-sulphonyl Chloride

M-Clarity™ Qualitätsstufe = MQ100

556505-1G
556505-1GEA 229 EUR
556505-1G
4-Chloro-7-chlorosulphonyl-2,1,3-benzoxadiazole 97%, Sigma-Aldrich®
4-Chloro-7-chlorosulphonyl-2,1,3-benzoxadiazole

Bifunctional fluorescent reagent. Cross-linking reagent.

Formel: C₆H₂Cl₂N₂O₃S
Molecular Weight: 253,05 g/mol
Schmelzpunkt: 86,..90 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00191405
CAS-Nummer: 142246-48-8
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a45fde5fde70c77442bfeab55373f5d82322bd8&vid=0a45fde5fde70c77442bfeab55373f5d82322bd8&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Off-White
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Titration by AgNO3 96.5 - 103.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR