Rubidium Standard for AAS, TraceCERT®, Supelco®
Lieferant: Merck
55727-100ML
55727-100MLEA
132
EUR
55727-100ML
Rubidium Standard for AAS, TraceCERT®, Supelco®
Standards
Kationischer Standards
Rubidium-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034