2-Amino-6-(trifluoromethoxy)benzothiazole ≥97,0% (durch HPLC), Calbiochem®
Lieferant: Merck Millipore Calbiochem
Synonyme:
6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole
557324-50
557324-50EA
186
EUR
557324-50
2-Amino-6-(trifluoromethoxy)benzothiazole ≥97,0% (durch HPLC), Calbiochem®
2-Amino-6-(trifluoromethoxy)benzothiazole
A cell-permeable benzothiazole compound that potently inhibits glutamate release and blocks Na+ channels. Shown to function as a polyglutamine aggregation inhibitor and stimulates the synthesis of NGF, BDNF, and GDNF in mouse astrocyte cultures. Reported to offer neuroprotection, slow down disease progression in ALS (amyotrophic lateral sclerosis) patients, and prolong the lifespan of animal models of HD (Huntington’s disease).
Formel:
C₈H₅F₃N₂OS Molecular Weight: 234,20 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00210213 CAS-Nummer: 1744-22-5 |
Spezifikation Testergebnisse
Form | White to off-white solid |
Purity | ≥97% by HPLC |
Contaminants | Heavy metals: ≤20 ppm |
Storage | +2 °C to +8 °C |
Stability | Stable for up to 3 months at –20 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...