Selen-L-methionin 97 HPLC_ASSAY_METHOD, Solid, Calbiochem®, Millipore®

Lieferant: Merck Millipore Calbiochem
Gefahr

Synonyme: L-SelenoMethionine, (S)-2-Amino-4-(methylseleno)butyric Acid, H-Se-Met-OH

MQ100

561505-250 561505-1
561505-250EA 300 EUR
561505-250 561505-1
Selen-L-methionin 97 HPLC_ASSAY_METHOD, Solid, Calbiochem®, Millipore®
Selen-L-methionin

It increases the expression of glutathione peroxidase.

Formel: C₅H₁₁NO₂Se
Molecular Weight: 196.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037210
CAS-Nummer: 3211-76-5
UN: 3283
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Synonyms SeMet
Form White to Off-white Solid
Purity ≥98% by HPLC
Contaminants (Heavy Metals) ≤10 ppm
Contaminants (Selenium) ≥39,5%
Ship Code Ambient Temperature only
Toxicity Carcinogenic / Teratogenic
Storage 15 to 30 °C
Do not freeze Ok to freeze
Special Instructions Unstable in solution; reconstitute just prior to use
Solubility H2O (50 mg/ml)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR