Organic reference standard, Rhodamine 6G, analytical standard, Supelco®
Lieferant: Merck
56226-25MG
56226-25MGEA 165 EUR
56226-25MG
Organic reference standard, Rhodamine 6G, analytical standard, Supelco®
Standards Organische Standards
A rhodamine analog useful in Pgp efflux assays. Has been used to characterize kinetics of MRP1- mediated efflux.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6649d5ed550432c785fcc5b7c45136b083a1fe21&vid=6649d5ed550432c785fcc5b7c45136b083a1fe21&prid=web&clid=SAPDAM