Cer(IV)sulfat 0.1 N, BAKER ANALYZED® volumetrische Lösung, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Cerdisulfat
5626-02EA
132.23
EUR
5626-02
Cer(IV)sulfat 0.1 N, BAKER ANALYZED® volumetrische Lösung, J.T.Baker®
Cer(IV)sulfat
Standardised at 25 °C against chemicals whose certification is traceable to NIST. Lot standardisation value, accurate to ±1 part per 1000, provided on label.
Formel:
Ce(SO₄)₂ Molecular Weight: 332,24 g/mol Schmelzpunkt: 195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00148852 CAS-Nummer: 13590-82-4 EINECS: 237-029-5 UN: 3264 ADR: 8,II Merck Index: 12,02036 |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | 0.095 - 0.105 |
Normality | 0.095 - 0.105 |
Standardization at 25°C traceable to NIST Standard Reference Material. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...