Ammoniumthiocyanat 0.1 N in wässriger Lösung, BAKER ANALYZED® volumetrische Lösung, J.T.Baker®
Lieferant: Avantor
Synonyme: Ammoniumsulfocyanid, Ammoniumrhodanid
5627-02EA 82.5 EUR
5627-02
Ammoniumthiocyanat 0.1 N in wässriger Lösung, BAKER ANALYZED® volumetrische Lösung, J.T.Baker®
Ammoniumthiocyanat
Standardised at 25 °C against chemicals whose certification is traceable to NIST. Lot standardisation value, accurate to ±1 part per 1000, provided on label.
Formel: NH₄SCN
Molecular Weight: 76,12 g/mol
Dichte: 1,10 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011428
CAS-Nummer: 1762-95-4
EINECS: 217-175-6
Bestellen
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use 0.095 - 0.105
Normality 0.095 - 0.105
Standardization at 25°C traceable to NIST Standard Reference Material.