Kaliumhydroxid 0.5 N in Methanol, BAKER ANALYZED® volumetrische Lösung, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Kalihydrat, Ätzkali
5631.1000EA
58.28
EUR
5631.1000
Kaliumhydroxid 0.5 N in Methanol, BAKER ANALYZED® volumetrische Lösung, J.T.Baker®
Kaliumhydroxid
Standardised at 25 °C against chemicals whose certification is traceable to NIST. Lot standardisation value, accurate to ±1 part per 1000, provided on label.
Formel:
KOH Molecular Weight: 56,11 g/mol Dichte: 0,80 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003553 CAS-Nummer: 1310-58-3 EINECS: 215-181-3 UN: 3286 ADR: 3,II |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | 0.497 - 0.503 |
Normality | 0.497 - 0.503 |
Chloride (Cl) | ≤10 ppm |
Nitrogen Compounds (as N) | ≤1 ppm |
ACS - Heavy Metals (as Pb) | ≤1 ppm |
Trace Impurities - Iron (Fe) | ≤0.5 ppm |
Standardization at 25°C traceable to NIST Standard Reference Material. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...