Nitrogen generator, Solaris
Lieferant: PEAK SCIENTIFIC
65-0001
564-0277EA 7000 EUR
564-0277
Nitrogen generator, Solaris
Gasgeneratoren Stickstoff-Generatoren
This nitrogen generator has been engineered and designed as a gas delivery solution for ELSD, Compact Mass Spectrometer instruments and TLD readers which can reduce downtime and increase workflow efficiencies.
  • Space saving design
  • Variable outlet flow, pressure and purity
  • Optional Solaris air compressor stackable module
  • Colour indicative LED lighting for easy status awareness
The generator can be placed on a bench top and paired with an additional air compressor unit to provide air supply for labs without an in-house air supply or those who wish to contain their gas supply in a single system.
Min./max. operating temperature: 5 to 35 °C
Max. pressure: 6,9 bar (user adjustable)
Gas outlet: 1×¼" BSPP
Standards: Manufactured and tested to highest spec in UK, CE/FCC certified.