Air compressor, corona
Lieferant: PEAK SCIENTIFIC
85-0555
564-0299EA 5460 EUR
564-0299
Air compressor, corona
Gasgeneratoren Druckluftstationen
The corona air compressor has been engineered to stack neatly with the corona 1010 nitrogen generator providing compressed air for laboratories without an external source of gas. The corona series has been developed to meet the requirements of the thermo scientific corona veo Charged aerosol detector.
  • Stackable with corona 1010 nitrogen generator
  • Provides air for corona system for laboratories without an external supply
  • Gas is supplied on demand so generator works to your schedule
  • 1 year comprehensive on-site warranty