L(+)-Serin, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
(S)-(+)-2-Amino-3-hydroxypropionsäure, (S)-(+)-Serin, H-Ser-OH
5650-100
5650-100EA
107
EUR
5650-100
L(+)-Serin, Millipore®
L(+)-Serin
Formel:
C₃H₇NO₃ Molecular Weight: 105,09 g/mol Siedepunkt: 259 °C (1013 hPa) Schmelzpunkt: 215…225 °C Dichte: 1,6 g/cm³ (22 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064224 CAS-Nummer: 56-45-1 EINECS: 200-274-3 Merck Index: 14,00846 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...