Compound E ≥95% (durch HPLC) Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem

Synonyme: (S)-2-(2-(3,5-Difluorophenyl)acetamido)-N-((S)-1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)propanamide

Verpackt unter Schutzgas

565790-500 565790-1MG
565790-500EA 323 EUR
565790-500 565790-1MG
Compound E ≥95% (durch HPLC) Calbiochem®, Sigma-Aldrich®
Compound E

A cell-permeable, potent, selective, peptidomimetic, non-transition-state analog inhibitor of γ-secretase and Notch processing (IC₅₀=300 pM for Aβ₄₀ in CHO cells overexpressing wild type βAPP; 240 pM for Aβ₄₀, 370 pM for Aβ₄₂, and 320 pM for NICD, respectively, in HEK293 cells stably transfected with βAPP₆₉₅
and mNotchδE; 100 pM for both Aβ₄₀ and Aβ₄₂ in SH-SY5Y cells stably transfected with SPA4CT).


Lowers Aβ levels in several APP transgenic mouse models. Reported to bind to presenilins and suppress the proteolytic cleavage of transmembrane protein substrates, including APLP1 and APLP2, CD44, ErbB4, E-cadherin, low density lipoprotein receptor-related proteins, Notch ligands, and p75NTR. Only weakly affects presenilinase activity at much higher concentrations (200 to 400 µM).

Formel: C₂₇H₂₄F₂N₄O₃
Lagertemperatur: Kühlschrank
MDL: MFCD04974531
CAS-Nummer: 209986-17-4

Bestellen

Spezifikation Testergebnisse

Purity (by HPLC) ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR