Natriumhydroxid 2.5 mol/l (2.5 N) in wässriger Lösung, BAKER ANALYZED®, Laborreagens volumetrische Lösung, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Caustic soda

5666-02EA 61.38 EUR
5666-02
Natriumhydroxid 2.5 mol/l (2.5 N) in wässriger Lösung, BAKER ANALYZED®, Laborreagens volumetrische Lösung, J.T.Baker®
Sodium hydroxide

J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications.

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 119…135 °C (1013 hPa)
Schmelzpunkt: 9…10 °C
Dichte: 1.53 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 2.40 - 2.60
Protect from air to avoid absorption of carbon dioxide. = 5 ppm
Normality 2.40 - 2.60
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR