Natriumhydroxid 5 N in wässriger Lösung, Biotechreagenz, J.T.Baker®

Lieferant: Avantor
Gefahr


Filtriert über 0,2 µm-Filter

5668-07EA 1230 EUR
5668-07 5668-02 5668-03 5668-09
Natriumhydroxid 5 N in wässriger Lösung, Biotechreagenz, J.T.Baker®
Sodium hydroxide

Made from USP purified water and NF (multicompendial) sodium hydroxide pellets that meet BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.

Formel: NaOH
Siedepunkt: 109 °C (1013 hPa)
Dichte: 1,17 kg/L
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Protect from air to avoid absorption of carbon dioxide.
CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless liquid) Passes Test
Normality 4.9 - 5.1
Chloride (Cl) ≤10 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Iron (Fe) ≤2 ppm
Made from U.S.P. Purified Water and Sodium Hydroxide pellets, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR