Sinefungin 95 HPLC_ASSAY_METHOD, InSolution™, Calbiochem®, Sigma-Aldrich®
Lieferant: Merck Millipore Calbiochem
Inert_Gas_filled
567051-2
567051-2EA
280
EUR
567051-2
Sinefungin 95 HPLC_ASSAY_METHOD, InSolution™, Calbiochem®, Sigma-Aldrich®
Sinefungin
An anti-leishmanial nucleoside antibiotic that acts as an S-adenosyl-L-methionine (SAM, AdoMet) methyltransferase-specific inhibitor.
Verpackung: Packaged under inert gas.
Lagertemperatur:
Gefrierschrank |
CAS-Nummer:
58944-73-3 |
Spezifikation Testergebnisse
Purity | ≥95% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...