Natriumbutyrat, Millipore®
Lieferant: Merck Millipore Calbiochem
567430-250
567430-250EA
84.7
EUR
567430-250
Natriumbutyrat, Millipore®
Natriumbutyrat
Formel:
C₄H₇NaO₂ Schmelzpunkt: 244 °C Dichte: 1,324 g/cm³ (30 °C) |
MDL:
MFCD00002816 CAS-Nummer: 156-54-7 EINECS: 205-857-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...