Nitroprussid-Natrium Dihydrat, Millipore®
Lieferant: Merck Millipore Calbiochem
|
Gefahr
|
Synonyme:
Dinatriumpentacyanonitrosylferrat(III) Dihydrat, Natriumnitrosopentacyanoferrat(III) Dihydrat, di-Natriumnitrosylpentacyanoferrat(III) Dihydrat, Natriumnitroprussiat Dihydrat, di-Natriumpentacyanonitrosylferrat(III) Dihydrat
567538-100
567538-100EA
69.6
EUR
567538-100
Nitroprussid-Natrium Dihydrat, Millipore®
Nitroprussid-Natrium Dihydrat
Formel:
Na₂[Fe(CN)₅NO]·2H₂O Molecular Weight: 297,95 g/mol Dichte: 1,72 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149192 CAS-Nummer: 13755-38-9 EINECS: 238-373-9 UN: 1588 ADR: 6.1,III Merck Index: 13,08722 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...