Natriumorthovanadat, Millipore®

Lieferant: Merck Millipore Calbiochem
Achtung

567540-5
567540-5EA 72.7 EUR
567540-5
Natriumorthovanadat, Millipore®
Natriumorthovanadat
Formel: Na₃O₄V
Molecular Weight: 183.91 g/mol
Schmelzpunkt: 850…866 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003511
CAS-Nummer: 13721-39-6
EINECS: 237-287-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR