Spermidin Trihydrochlorid, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: 4-Azaoctamethylendiamin Trihydrochlorid, N-(3-Aminopropyl)-1,4-diaminobutan Trihydrochlorid, 4-Azoniaoctamethylendiammoniumtrichlorid

56766-5
56766-5EA 280 EUR
56766-5
Spermidin Trihydrochlorid, Millipore®
Spermidin Trihydrochlorid
Formel: C₇H₂₂Cl₃N₃
Molecular Weight: 254,63 g/mol
Schmelzpunkt: 256…258 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00012918
CAS-Nummer: 334-50-9
EINECS: 206-379-0
Merck Index: 12,08893

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR