Spermin Tetrahydrochlorid, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine) tetrahydrochloride

5677-5
5677-5EA 336 EUR
5677-5
Spermin Tetrahydrochlorid, Millipore®
Spermin Tetrahydrochlorid
Formel: C₁₀H₃₀Cl₄N₄
MDL: MFCD00012914
CAS-Nummer: 306-67-2
EINECS: 206-189-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR