Kapillarsäulen, BP20 (Wax)
Lieferant: Trajan Scientific and Medical
054405 054421 054422 054424 054425 054427 054428 054431 054433 054434 054436 054437 054440 054442 054443 054448 054450 054451 0544515 054456 054462 054415 054438 054439 054444 054445 054457 054458
568-0145EA 395.2 EUR
568-0145 558-0324 558-0325 568-0147 568-0148 558-0253 558-0327 568-0150 558-0328 568-0152 558-0329 568-0153 568-0156 568-0157 568-0158 568-0162 568-0163 558-0330 568-0164 568-0166 568-0169 558-0323 568-0154 568-0155 568-0159 568-0160 568-0167 568-0168
Kapillarsäulen, BP20 (Wax)
Chromatographiesäulen
These polyethylene glycol columns are suitable for the analysis of alcohol, free acids, fatty acid methyl esters, aromatics, solvents, essential oils.
  • Polar phase
  • Cross-linked for stability and washing
Geeigneter Ersatz für: DB-Wax, HP-20M, Supelcowax 10, CB-Wax, Stabilwax, Carbowax, HP-Innowax, Rtx-WAX, PE-WAX, RH-WAX, ZB-WAX, TRWAX.
Max. Temperatur: 0,1–1,0 µm Filmdicke = 20 °C bis 260/280 °C; >1,0 µm Filmdicke = 20 °C bis 240/260 °C