SGE HT5 GC Capillary Columns
Lieferant: Trajan Scientific and Medical
054631 054632 054641 054642 054655 054657 054658 054667 054668 054670 054672 054633 054634 054640 054638
568-0182EA 492.9 EUR
568-0182 568-0183 568-0188 568-0189 568-0193 568-0194 568-0195 568-0199 568-0200 568-0201 568-0203 568-0184 568-0185 568-0001 SGEA054638
SGE HT5 GC Capillary Columns
Chromatographiesäulen
The SGE HT5 GC capillary column provides a great low-polarity option for GC analyses. Recommended for simulated distillation, general hydrocarbon profiles, pesticides/herbicides, GCMS applications.
  • Ultra high temperature columns
  • Unique phase – no equivalent phases
  • Ideal for simulated distillation applications (petroleum industry)
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.