Spanner, open end, SGE
Lieferant: SGE Analytical Science
18500002
568-0686EA 29 EUR
568-0686
Spanner, open end, SGE
Schraubenschlüssel
Spanner sizes and uses: 3/16" for use with mini unions, 1/4" for use with 1/16" nuts, 5/16" for use with micro control valves.