Industrial label printers with Brady workstation LAB suite, BradyPrinter i7100
Lieferant: Brady
306883 306886 306889 306880
568-0897EA 2292 EUR
568-0897 568-0899 568-0901 568-0895
Industrial label printers with Brady workstation LAB suite, BradyPrinter i7100
Drucker Label Printers
The BradyPrinter i7100 industrial thermal transfer bench top printer is ideal for customers requiring reliable high volume printing on lots of different materials within the aerospace, defence and mass transit, electronics, automotive, logistics, electrical, voice-datacom and lab markets.
  • 300 or 600 dpi
  • Applications include cable and wire identification, circuit board and component identification, and general and industrial identification
  • Mono colour
It guarantees accurate printing on sleeves, tags, labels, tapes, cable markers and much more. The printer comes with the Brady Workstation Product & Wire Identification software suite enabling you to immediately start designing professional labels. This suite combines frequently used software apps to create product and cable identification labels.
Barcode types include Code 39; Code 93; Code 39 Full ASCII; code 128 A; code 128 B; code 128 C; EAN 8; EAN13; EAN/UCC 128/GS1-128; EAN/UPC; FIM; HIBC; Interleaved 2/5; ident and lead code of Deutsche Post AG; Codabar; JAN8; JAN 13; MSI; Plessey; Postnet; RSS14; UPC A; UPC E; UPC E0; Aztec; Codablock F; DataMatrix; PDF417; Micro PDF417; UPS MaxiCode; QR code; RSS 14 truncated, limited stacked and stacked omnidirectional; EAN/GS1 DataMatrix; GS1 DataBar.
Languages: Arabic (Egypt); Bulgarian; Croatian; Czech; Danish; Dutch; English; Finnish; French; German; Greek; Hungarian; Italian; Lithuanian; Macedonian; Norwegian (Bokmål); Persian (Farsi); Polish; Portuguese; Romanian; Russian; Serbian; Slovenian; Spanish; Swedish and Turkish.
Zubehörinformationen: Available hardware options include Cutter with Tray, Cutter Perforator, Present Sensor, Peel Off, Rewinder and Platen Rollers; contact your local Avantor representative for more information.
Lieferumfang: This i7100 printer comes in a cardboard box. It ships with an EU or UK power cord, 2 empty ribbon cores, a USB cable, a product CD holding the driver, the user manual in languages, the configuration/settings manual in English, a printed user manual in English and Brady Workstation Laboratory ID Suite. The Peel and Present module is optionally pre-installed.