Immersion oil for microscopy, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
56822-1L
56822-250ML
56822-50ML
56822-1LEA
207
EUR
56822-1L
SIAM56822-250ML
56822-50ML
Immersion oil for microscopy, Sigma-Aldrich®
Färbemittel
Mikroskopie- und Färbereagenzien
Immersion oil is used for high resolution (1000X) light microscopy work under oil immersion objective lens.
- Suitable for Hematology
- Suitable for Histology