Staurosporine (aus Streptomyces species), InSolution™, Millipore®

Lieferant: Merck Millipore Calbiochem

569396-100
569396-100EA 155 EUR
569396-100
Staurosporine (aus Streptomyces species), InSolution™, Millipore®
Staurosporine

InSolution™ Staurosporine, Streptomyces sp. is provided as a 1 mM (100 µg/214 µl) solution of Staurosporine in DMSO.

Formel: C₂₈H₂₆N₄O₃
MDL: MFCD18252446
CAS-Nummer: 62996-74-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR