Blei(II)iodid ≥98,5%

Lieferant: Thermo Fisher Scientific
Gefahr

57103.18 57103.30
57103.18EA 76.9 EUR
57103.18 57103.30
Blei(II)iodid ≥98,5%
Blei(II)iodid

Purity: 98.5% CAS No.: 10101-63-0 Molecular Formula: I2Pb Formula Weight: 461.00 Physical Form: Powder Density: 6.16 Appearance: Yellow Odour: Odourless Melting Point: 402° Boiling Point: 954° Solubility: Partially soluble in water. Freely soluble in sodium thiosulfate solution. Soluble in concentrated solutions of alkali iodides. Insoluble in alcohol and cold HCl Merck Reference: 14,5411 UN No.: 2291 MDL No.: MFCD00011163

Formel: I₂Pb
Siedepunkt: 954 °C
Schmelzpunkt: 402 °C
Dichte: 6,16
Lagertemperatur: Raumtemperatur
MDL: MFCD00011163
CAS-Nummer: 10101-63-0
EINECS: 233-256-9
UN: 2291
ADR: 6.1,III
Merck Index: 14,5411

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR