N-[3-Cyano-3-[4-(dicyanomethyl)phenyl]-2-propenylidene]-N-ethyl-ethaniminium inner salt, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
572136-50MG
572136-50MGEA
157
EUR
572136-50MG
N-[3-Cyano-3-[4-(dicyanomethyl)phenyl]-2-propenylidene]-N-ethyl-ethaniminium inner salt, Sigma-Aldrich®
N-[3-Cyano-3-[4-(dicyanomethyl)phenyl]-2-propenylidene]-N-ethyl-ethaniminium inner salt
Formel:
C₁₇H₁₆N₄ Molecular Weight: 276,34 g/mol Lagertemperatur: Room Temperature |
MDL:
MFCD04040956 CAS-Nummer: 174280-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...