SATA (N-Succinimidyl S-Acetylthioglycolate) ≥98% (durch HPLC), weißer Feststoff, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: N-Succinimidyl-S-acetylthioacetate, S-Acetylthioglycolic Acid N-Succinimidyl Ester, SATA, 2,5-Dioxopyrrolidin-1-yl 2-(acetylthio)acetate

573100-100
573100-100EA 177 EUR
573100-100
SATA (N-Succinimidyl S-Acetylthioglycolate) ≥98% (durch HPLC), weißer Feststoff, Millipore®
SATA (N-Succinimidyl S-Acetylthioglycolate)

A useful agent for introducing thiol groups, in protected form, into various proteins. Reacts with primary amines to add protected sulfhydryl groups. When treated with hydroxylamine, yields a free sulfhydryl group that can be conjugated to maleimide-modified proteins.


Synonyms: N-Succinimidyl-S-acetylthioacetate

Formel: C₈H₉NO₅S
Molecular Weight: 231,22 g/mol
MDL: MFCD00036891
CAS-Nummer: 76931-93-6

Bestellen

Spezifikation Testergebnisse

Form White solid
Purity ≥98% by HPLC
Storage –20 °C
Stability Stable for up to 3 months at –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR