SynaptoRed™ Reagens ≥95% (durch HPLC), Calbiochem®, Sigma-Aldrich®
Lieferant: Merck Millipore Calbiochem
Synonyme:
FM® 4-64, Pyridinium, 4-[6-[4-(diethylamino)phenyl]- 1,3,5-hexatrienyl]-1-[3-(triethylammonio) propyl]-, dibromide, SynaptoRed™, MM 4-64
M-Clarity™ Qualitätsstufe = MQ100
A lipophilic, water-soluble styrylpyridinium red fluoroprobe that is useful for staining active recycling synaptic vesicles.
Also useful in studying endocytosis and identifying cell membrane boundaries. The compound is non-fluorescent in aqueous solution but becomes fluorescent when incorporated into the plasma membrane. Reversible, photostable, and does not diffuse out of internalized vesicles. Can also be used in combination with other green fluoroprobes for double-labeling experiments.
A lipophilic, nontoxic, water-soluble styrylpyridinium red fluoroprobe that is useful for staining active recycling synaptic vesicles. Also useful for studying endocytosis, membrane turnover, and identifying cell membrane boundaries. This compound is non-fluorescent in aqueous solution, but becomes fluorescent when incorporated into the plasma membrane. Reversible, photostable material that does not diffuse out of internalized vesicles. Can also be used in combination with the SynaptoGreen™ (Cat. No. 574798) to image plasma membranes independently in two colors. Excitation max.: 538-548 nm; emission max.: ~735 nm.
SynaptoRed is a trademark of Biotium, Inc.
Verpackung: Packaged under inert gas
Formel:
C₃₀H₄₅Br₂N₃ Molecular Weight: 607,51 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00467819 CAS-Nummer: 162112-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...