Acetic Acid : Triethylamine solution 1:1 2M:2M in wässriger Lösung, LiChropur™ geeignet für HPLC, Supelco®
Lieferant: Merck
Synonyme:
Acetic acid – TEA solution, Triethyl Ammonium Acetate
57619-100ML
57619-100MLEA
207
EUR
57619-100ML
Acetic Acid : Triethylamine solution 1:1 2M:2M in wässriger Lösung, LiChropur™ geeignet für HPLC, Supelco®
Acetic Acid : Triethylamine solution
Formel:
CH₃COOH·N(CH₂CH₃)₃ Molecular Weight: 161,24 g/mol |
MDL:
MFCD00151725 CAS-Nummer: 5204-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...